2-Amino-N-(tetrahydro-2H-pyran-4-ylmethyl)acetamide hydrochloride

Supplier: Matrix Scientific

053547-500MG 053547-2.5G
101904-074EA 364.62 USD
101904-074 101904-076
2-Amino-N-(tetrahydro-2H-pyran-4-ylmethyl)acetamide hydrochloride
2-Amino-N-(tetrahydro-2H-pyran-4-ylmethyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₇ClN₂O₂
MDL Number: MFCD13562864
CAS Number: 1219976-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR