6-Chloro-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-pyridinamine

Supplier: Matrix Scientific

053546-500MG 053546-2.5G
101904-070EA 364.62 USD
101904-070 101904-072
6-Chloro-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-pyridinamine
6-Chloro-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O
MDL Number: MFCD13562863
CAS Number: 1219967-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR