2-Chloro-N-(tetrahydro-2H-pyran-4-ylmethyl)nicotinamide

Supplier: Matrix Scientific

053543-500MG 053543-2.5G
101904-058EA 364.62 USD
101904-058 101904-060
2-Chloro-N-(tetrahydro-2H-pyran-4-ylmethyl)nicotinamide
2-Chloro-N-(tetrahydro-2H-pyran-4-ylmethyl)nicotinamide

Hazard: Irritant

Formula: C₁₂H₁₅ClN₂O₂
MW: 254.72 g/mol
MDL Number: MFCD13562860
CAS Number: 1220035-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR