N2-(Tetrahydro-2H-pyran-4-ylmethyl)-2,3-pyridinediamine

Supplier: Matrix Scientific

053542-500MG 053542-2.5G
101904-054EA 364.62 USD
101904-054 101904-056
N2-(Tetrahydro-2H-pyran-4-ylmethyl)-2,3-pyridinediamine
N2-(Tetrahydro-2H-pyran-4-ylmethyl)-2,3-pyridinediamine

Hazard: Irritant

Formula: C₁₁H₁₇N₃O
MDL Number: MFCD13562859
CAS Number: 1219976-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR