2-[(Tetrahydro-2H-pyran-4-ylmethyl)amino]isonicotinic acid
Supplier: Matrix Scientific
053540-500MG
053540-2.5G
101904-046EA
364.62
USD
101904-046
101904-048
2-[(Tetrahydro-2H-pyran-4-ylmethyl)amino]isonicotinic acid
2-[(Tetrahydro-2H-pyran-4-ylmethyl)amino]isonicotinic acid
Hazard: Irritant
Formula:
C₁₂H₁₆N₂O₃ MW: 236.27 g/mol |
MDL Number:
MFCD13562857 CAS Number: 1219960-34-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...