6-Chloro-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-pyrazinamine

Supplier: Matrix Scientific

053539-500MG 053539-2.5G
101904-042EA 364.62 USD
101904-042 101904-044
6-Chloro-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-pyrazinamine
6-Chloro-N-(tetrahydro-2H-pyran-4-ylmethyl)-2-pyrazinamine

Hazard: Irritant

Formula: C₁₀H₁₄ClN₃O
MDL Number: MFCD13562856
CAS Number: 1220029-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR