3-Nitro-4-[(tetrahydro-2H-pyran-4-ylmethyl)amino]benzoic acid

Supplier: Matrix Scientific

053535-500MG 053535-2.5G
101904-026EA 364.62 USD
101904-026 101904-028
3-Nitro-4-[(tetrahydro-2H-pyran-4-ylmethyl)amino]benzoic acid
3-Nitro-4-[(tetrahydro-2H-pyran-4-ylmethyl)amino]benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₅
MDL Number: MFCD13562852
CAS Number: 1220018-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR