N-(4-Methoxyphenyl)-4-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
Synonyms:
N-(4-Methoxyphenyl)piperidine-4-carboxamide hydrochloride
053531-500MG
053531-2.5G
101904-010EA
364.62
USD
101904-010
101904-012
N-(4-Methoxyphenyl)-4-piperidinecarboxamide hydrochloride
N-(4-Methoxyphenyl)-4-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₉ClN₂O₂ MW: 270.76 g/mol |
MDL Number:
MFCD13562849 CAS Number: 1186049-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...