N-(4-Methoxyphenyl)-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053529-500MG 053529-2.5G
101904-002EA 364.62 USD
101904-002 101904-004
N-(4-Methoxyphenyl)-2-piperidinecarboxamide hydrochloride
N-(4-Methoxyphenyl)-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₉ClN₂O₂
MDL Number: MFCD13562847
CAS Number: 1236264-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR