N-(4-Methoxyphenyl)-2-pyrrolidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053528-500MG
053528-2.5G
101903-998EA
364.62
USD
101903-998
101904-000
N-(4-Methoxyphenyl)-2-pyrrolidinecarboxamide hydrochloride
N-(4-Methoxyphenyl)-2-pyrrolidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₇ClN₂O₂ |
MDL Number:
MFCD11505474 CAS Number: 1219404-87-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...