2-Amino-N-(4-methoxyphenyl)-3-methylbutanamide hydrochloride
Supplier: Matrix Scientific
053525-500MG
053525-2.5G
101903-986EA
364.62
USD
101903-986
101903-988
2-Amino-N-(4-methoxyphenyl)-3-methylbutanamide hydrochloride
2-Amino-N-(4-methoxyphenyl)-3-methylbutanamide hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₉ClN₂O₂ |
MDL Number:
MFCD13562844 CAS Number: 1236255-24-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...