2-Amino-N-(4-methoxyphenyl)propanamide hydrochloride

Supplier: Matrix Scientific

053522-500MG 053522-2.5G
101903-974EA 364.62 USD
101903-974 101903-976
2-Amino-N-(4-methoxyphenyl)propanamide hydrochloride
2-Amino-N-(4-methoxyphenyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O₂
MDL Number: MFCD13562842
CAS Number: 1236256-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR