2-Amino-N-(4-methoxyphenyl)acetamide hydrochloride
Supplier: Matrix Scientific
053521-500MG
053521-2.5G
101903-970EA
364.62
USD
101903-970
101903-972
2-Amino-N-(4-methoxyphenyl)acetamide hydrochloride
2-Amino-N-(4-methoxyphenyl)acetamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₃ClN₂O₂ |
MDL Number:
MFCD13562841 CAS Number: 85622-19-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...