6-Chloro-N-(4-methoxyphenyl)-2-pyridinamine

Supplier: Matrix Scientific

053520-500MG 053520-2.5G
101903-966EA 364.62 USD
101903-966 101903-968
6-Chloro-N-(4-methoxyphenyl)-2-pyridinamine
6-Chloro-N-(4-methoxyphenyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₁₁ClN₂O
MDL Number: MFCD13562840
CAS Number: 1219976-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR