5-Bromo-N-(4-methoxyphenyl)-3-methyl-2-pyridinamine

Supplier: Matrix Scientific

053518-500MG 053518-2.5G
101903-958EA 364.62 USD
101903-958 101903-960
5-Bromo-N-(4-methoxyphenyl)-3-methyl-2-pyridinamine
5-Bromo-N-(4-methoxyphenyl)-3-methyl-2-pyridinamine

Hazard: Irritant

Formula: C₁₃H₁₃BrN₂O
MDL Number: MFCD13562838
CAS Number: 1219967-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR