2-Chloro-N-(4-methoxyphenyl)nicotinamide

Supplier: Matrix Scientific

053517-500MG 053517-2.5G
101903-954EA 364.62 USD
101903-954 101903-956
2-Chloro-N-(4-methoxyphenyl)nicotinamide
2-Chloro-N-(4-methoxyphenyl)nicotinamide

Hazard: Irritant

Formula: C₁₃H₁₁ClN₂O₂
MDL Number: MFCD00665154
CAS Number: 56149-30-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR