N2-(4-Methoxyphenyl)-2,3-pyridinediamine

Supplier: Matrix Scientific

053516-500MG 053516-2.5G
101903-950EA 364.62 USD
101903-950 101903-952
N2-(4-Methoxyphenyl)-2,3-pyridinediamine
N2-(4-Methoxyphenyl)-2,3-pyridinediamine

Hazard: Irritant

Formula: C₁₂H₁₃N₃O
MDL Number: MFCD09757487
CAS Number: 41010-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR