2-(4-Methoxyanilino)isonicotinic acid

Supplier: Matrix Scientific

053515-500MG 053515-2.5G
101903-946EA 364.62 USD
101903-946 101903-948
2-(4-Methoxyanilino)isonicotinic acid
2-(4-Methoxyanilino)isonicotinic acid

Hazard: Irritant

Formula: C₁₃H₁₂N₂O₃
MDL Number: MFCD09414717
CAS Number: 85726-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR