6-Chloro-N-(4-methoxyphenyl)-2-pyrazinamine

Supplier: Matrix Scientific

053514-500MG 053514-2.5G
101903-942EA 364.62 USD
101903-942 101903-944
6-Chloro-N-(4-methoxyphenyl)-2-pyrazinamine
6-Chloro-N-(4-methoxyphenyl)-2-pyrazinamine

Hazard: Irritant

Formula: C₁₁H₁₀ClN₃O
MW: 235.67 g/mol
MDL Number: MFCD13562837
CAS Number: 874143-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR