3-Chloro-N-(4-methoxyphenyl)-5-(trifluoromethyl)-2-pyridinamine

Supplier: Matrix Scientific

053513-500MG 053513-2.5G
101903-938EA 364.62 USD
101903-938 101903-940
3-Chloro-N-(4-methoxyphenyl)-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N-(4-methoxyphenyl)-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₃H₁₀ClF₃N₂O
MDL Number: MFCD13562836
CAS Number: 581792-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR