5-Bromo-N-(4-methoxyphenyl)-4-methyl-2-pyridinamine
Supplier: Matrix Scientific
053511-500MG
053511-2.5G
101903-930EA
364.62
USD
101903-930
101903-932
5-Bromo-N-(4-methoxyphenyl)-4-methyl-2-pyridinamine
5-Bromo-N-(4-methoxyphenyl)-4-methyl-2-pyridinamine
Hazard: Irritant
Formula:
C₁₃H₁₃BrN₂O |
MDL Number:
MFCD13562834 CAS Number: 1220020-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...