4-(4-Methoxyanilino)-3-nitrobenzoic acid
Supplier: Matrix Scientific
053510-500MG
053510-2.5G
101903-926EA
364.62
USD
101903-926
101903-928
4-(4-Methoxyanilino)-3-nitrobenzoic acid
4-(4-Methoxyanilino)-3-nitrobenzoic acid
Hazard: Irritant
Formula:
C₁₄H₁₂N₂O₅ |
MDL Number:
MFCD00418447 CAS Number: 148304-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...