Ethyl-3-amino-4-(4-methoxyanilino)benzoate

Supplier: Matrix Scientific

053509-500MG 053509-2.5G
101903-922EA 364.62 USD
101903-922 101903-924
Ethyl-3-amino-4-(4-methoxyanilino)benzoate
Ethyl-3-amino-4-(4-methoxyanilino)benzoate

Hazard: Irritant

Formula: C₁₆H₁₈N₂O₃
MDL Number: MFCD13562833
CAS Number: 1219960-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR