3-Amino-4-chloro-N-(4-fluorophenyl)benzenesulfonamide
Supplier: Matrix Scientific
053508-500MG
053508-2.5G
101903-918EA
364.62
USD
101903-918
101903-920
3-Amino-4-chloro-N-(4-fluorophenyl)benzenesulfonamide
3-Amino-4-chloro-N-(4-fluorophenyl)benzenesulfonamide
Hazard: Irritant
Formula:
C₁₂H₁₀ClFN₂O₂S MW: 300.74 g/mol |
MDL Number:
MFCD12737294 CAS Number: 1041527-31-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...