N-(4-Fluorophenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
Supplier: Matrix Scientific
053507-500MG
053507-2.5G
101903-914EA
364.62
USD
101903-914
101903-916
N-(4-Fluorophenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
N-(4-Fluorophenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[4,3-c]pyridine-3-carboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₃H₁₄ClFN₄O MW: 296.73 g/mol |
MDL Number:
MFCD13562832 CAS Number: 1219967-22-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...