N-(4-Fluorophenyl)-3-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053505-500MG
053505-2.5G
101903-906EA
364.62
USD
101903-906
101903-908
N-(4-Fluorophenyl)-3-piperidinecarboxamide hydrochloride
N-(4-Fluorophenyl)-3-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₆ClFN₂O |
MDL Number:
MFCD13562830 CAS Number: 1219976-72-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...