N-(4-Fluorophenyl)-2-piperidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053504-500MG 053504-2.5G
101903-902EA 364.62 USD
101903-902 101903-904
N-(4-Fluorophenyl)-2-piperidinecarboxamide hydrochloride
N-(4-Fluorophenyl)-2-piperidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₆ClFN₂O
MDL Number: MFCD13562829
CAS Number: 1236262-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR