N-(4-Fluorophenyl)-2-pyrrolidinecarboxamide hydrochloride

Supplier: Matrix Scientific

053503-500MG 053503-2.5G
101903-898EA 364.62 USD
101903-898 101903-900
N-(4-Fluorophenyl)-2-pyrrolidinecarboxamide hydrochloride
N-(4-Fluorophenyl)-2-pyrrolidinecarboxamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₄ClFN₂O
MDL Number: MFCD13562828
CAS Number: 1236267-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR