2-Amino-N-(4-fluorophenyl)-3-phenylpropanamide hydrochloride
Supplier: Matrix Scientific
053502-500MG
053502-2.5G
101903-894EA
364.62
USD
101903-894
101903-896
2-Amino-N-(4-fluorophenyl)-3-phenylpropanamide hydrochloride
2-Amino-N-(4-fluorophenyl)-3-phenylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₁₅H₁₆ClFN₂O |
MDL Number:
MFCD13562827 CAS Number: 1236261-36-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...