2-Amino-N-(4-fluorophenyl)-2-methylpropanamide hydrochloride
Supplier: Matrix Scientific
053501-500MG
053501-2.5G
101903-890EA
364.62
USD
101903-890
101903-892
2-Amino-N-(4-fluorophenyl)-2-methylpropanamide hydrochloride
2-Amino-N-(4-fluorophenyl)-2-methylpropanamide hydrochloride
Hazard: Irritant
Formula:
C₁₀H₁₄ClFN₂O |
MDL Number:
MFCD13562826 CAS Number: 1219976-76-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...