2-Amino-N-(4-fluorophenyl)-3-methylbutanamide hydrochloride

Supplier: Matrix Scientific

053500-500MG 053500-2.5G
101903-886EA 364.62 USD
101903-886 101903-888
2-Amino-N-(4-fluorophenyl)-3-methylbutanamide hydrochloride
2-Amino-N-(4-fluorophenyl)-3-methylbutanamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClFN₂O
MDL Number: MFCD13562825
CAS Number: 1236255-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR