2-Amino-N-(4-fluorophenyl)propanamide hydrochloride
Supplier: Matrix Scientific
053497-500MG
053497-2.5G
101903-874EA
364.62
USD
101903-874
101903-876
2-Amino-N-(4-fluorophenyl)propanamide hydrochloride
2-Amino-N-(4-fluorophenyl)propanamide hydrochloride
Hazard: Irritant
Formula:
C₉H₁₂ClFN₂O |
MDL Number:
MFCD13562824 CAS Number: 1246172-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...