2-Amino-N-(4-fluorophenyl)acetamide hydrochloride

Supplier: Matrix Scientific

053496-500MG 053496-2.5G
101903-870EA 364.62 USD
101903-870 101903-872
2-Amino-N-(4-fluorophenyl)acetamide hydrochloride
2-Amino-N-(4-fluorophenyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₈H₁₀ClFN₂O
MDL Number: MFCD13562823
CAS Number: 137895-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR