6-Chloro-N-(4-fluorophenyl)-2-pyridinamine

Supplier: Matrix Scientific

053495-500MG 053495-2.5G
101903-866EA 364.62 USD
101903-866 101903-868
6-Chloro-N-(4-fluorophenyl)-2-pyridinamine
6-Chloro-N-(4-fluorophenyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₁H₈ClFN₂
MDL Number: MFCD13562822
CAS Number: 881419-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR