N-(5-Bromo-2-pyridinyl)-N-(4-fluorophenyl)amine

Supplier: Matrix Scientific

053494-500MG 053494-2.5G
101903-862EA 364.62 USD
101903-862 101903-864
N-(5-Bromo-2-pyridinyl)-N-(4-fluorophenyl)amine
N-(5-Bromo-2-pyridinyl)-N-(4-fluorophenyl)amine

Hazard: Irritant

Formula: C₁₁H₈BrFN₂
MDL Number: MFCD13562821
CAS Number: 918305-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR