N-(5-Bromo-3-methyl-2-pyridinyl)-N-(4-fluorophenyl)amine
Supplier: Matrix Scientific
053493-500MG
053493-2.5G
101903-858EA
364.62
USD
101903-858
101903-860
N-(5-Bromo-3-methyl-2-pyridinyl)-N-(4-fluorophenyl)amine
N-(5-Bromo-3-methyl-2-pyridinyl)-N-(4-fluorophenyl)amine
Hazard: Irritant
Formula:
C₁₂H₁₀BrFN₂ |
MDL Number:
MFCD13562820 CAS Number: 1219967-14-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...