6-Chloro-N-(4-fluorophenyl)-2-pyrazinamine
Supplier: Matrix Scientific
053488-500MG
053488-2.5G
101903-838EA
364.62
USD
101903-838
101903-840
6-Chloro-N-(4-fluorophenyl)-2-pyrazinamine
6-Chloro-N-(4-fluorophenyl)-2-pyrazinamine
Hazard: Irritant
Formula:
C₁₀H₇ClFN₃ |
MDL Number:
MFCD13562819 CAS Number: 642459-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...