3-Chloro-N-(4-fluorophenyl)-5-(trifluoromethyl)-2-pyridinamine

Supplier: Matrix Scientific

053487-500MG 053487-2.5G
101903-834EA 364.62 USD
101903-834 101903-836
3-Chloro-N-(4-fluorophenyl)-5-(trifluoromethyl)-2-pyridinamine
3-Chloro-N-(4-fluorophenyl)-5-(trifluoromethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₂H₇ClF₄N₂
MDL Number: MFCD13562818
CAS Number: 1219976-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR