N-(5-Bromo-4-methyl-2-pyridinyl)-N-(4-fluorophenyl)amine

Supplier: Matrix Scientific

053485-500MG 053485-2.5G
101903-826EA 364.62 USD
101903-826 101903-828
N-(5-Bromo-4-methyl-2-pyridinyl)-N-(4-fluorophenyl)amine
N-(5-Bromo-4-methyl-2-pyridinyl)-N-(4-fluorophenyl)amine

Hazard: Irritant

Formula: C₁₂H₁₀BrFN₂
MDL Number: MFCD13562817
CAS Number: 1220038-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR