Ethyl-3-amino-4-(4-fluoroanilino)benzoate
Supplier: Matrix Scientific
053483-500MG
053483-2.5G
101903-818EA
364.62
USD
101903-818
101903-820
Ethyl-3-amino-4-(4-fluoroanilino)benzoate
Ethyl-3-amino-4-(4-fluoroanilino)benzoate
Hazard: Irritant
Formula:
C₁₅H₁₅FN₂O₂ |
MDL Number:
MFCD13562816 CAS Number: 1219960-30-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...