N-(4-Pyridinylmethyl)-3-piperidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053479-500MG
053479-2.5G
101903-802EA
364.62
USD
101903-802
101903-804
N-(4-Pyridinylmethyl)-3-piperidinecarboxamide hydrochloride
N-(4-Pyridinylmethyl)-3-piperidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₈ClN₃O |
MDL Number:
MFCD13562813 CAS Number: 1220035-43-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...