N-(4-Pyridinylmethyl)-2-pyrrolidinecarboxamide hydrochloride
Supplier: Matrix Scientific
053477-500MG
053477-2.5G
101903-794EA
364.62
USD
101903-794
101903-796
N-(4-Pyridinylmethyl)-2-pyrrolidinecarboxamide hydrochloride
N-(4-Pyridinylmethyl)-2-pyrrolidinecarboxamide hydrochloride
Hazard: Irritant
Formula:
C₁₁H₁₆ClN₃O |
MDL Number:
MFCD13562811 CAS Number: 1236262-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...