2-Amino-3-phenyl-N-(4-pyridinylmethyl)propanamide hydrochloride

Supplier: Matrix Scientific

053476-500MG 053476-2.5G
101903-790EA 364.62 USD
101903-790 101903-792
2-Amino-3-phenyl-N-(4-pyridinylmethyl)propanamide hydrochloride
2-Amino-3-phenyl-N-(4-pyridinylmethyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₁₅H₁₈ClN₃O
MDL Number: MFCD13562810
CAS Number: 1236261-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR