2-Amino-3-methyl-N-(4-pyridinylmethyl)butanamide hydrochloride

Supplier: Matrix Scientific

053474-500MG 053474-2.5G
101903-782EA 364.62 USD
101903-782 101903-784
2-Amino-3-methyl-N-(4-pyridinylmethyl)butanamide hydrochloride
2-Amino-3-methyl-N-(4-pyridinylmethyl)butanamide hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₈ClN₃O
MDL Number: MFCD13562808
CAS Number: 1236264-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR