3-Amino-N-(4-pyridinylmethyl)propanamide hydrochloride

Supplier: Matrix Scientific

053473-500MG 053473-2.5G
101903-778EA 364.62 USD
101903-778 101903-780
3-Amino-N-(4-pyridinylmethyl)propanamide hydrochloride
3-Amino-N-(4-pyridinylmethyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₄ClN₃O
MDL Number: MFCD13562807
CAS Number: 1219957-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR