2-(Methylamino)-N-(4-pyridinylmethyl)acetamide hydrochloride

Supplier: Matrix Scientific

053472-500MG 053472-2.5G
101903-774EA 364.62 USD
101903-774 101903-776
2-(Methylamino)-N-(4-pyridinylmethyl)acetamide hydrochloride
2-(Methylamino)-N-(4-pyridinylmethyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₄ClN₃O
MDL Number: MFCD13562806
CAS Number: 1219964-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR