2-Amino-N-(4-pyridinylmethyl)propanamide hydrochloride

Supplier: Matrix Scientific

053471-500MG 053471-2.5G
101903-770EA 364.62 USD
101903-770 101903-772
2-Amino-N-(4-pyridinylmethyl)propanamide hydrochloride
2-Amino-N-(4-pyridinylmethyl)propanamide hydrochloride

Hazard: Irritant

Formula: C₉H₁₄ClN₃O
MDL Number: MFCD13562805
CAS Number: 1236263-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR