2-Amino-N-(4-pyridinylmethyl)acetamide hydrochloride
Supplier: Matrix Scientific
053470-500MG
053470-2.5G
101903-766EA
364.62
USD
101903-766
101903-768
2-Amino-N-(4-pyridinylmethyl)acetamide hydrochloride
2-Amino-N-(4-pyridinylmethyl)acetamide hydrochloride
Hazard: Irritant
Formula:
C₈H₁₂ClN₃O |
MDL Number:
MFCD13562804 CAS Number: 1220018-62-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...