6-Chloro-N-(4-pyridinylmethyl)-2-pyridinamine

Supplier: Matrix Scientific

053469-500MG 053469-2.5G
101903-762EA 364.62 USD
101903-762 101903-764
6-Chloro-N-(4-pyridinylmethyl)-2-pyridinamine
6-Chloro-N-(4-pyridinylmethyl)-2-pyridinamine

Hazard: Irritant

Formula: C₁₁H₁₀ClN₃
MDL Number: MFCD13562803
CAS Number: 1220018-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR