5-Bromo-N-(4-pyridinylmethyl)-2-pyridinamine
Supplier: Matrix Scientific
053468-500MG
053468-2.5G
101903-758EA
364.62
USD
101903-758
101903-760
5-Bromo-N-(4-pyridinylmethyl)-2-pyridinamine
5-Bromo-N-(4-pyridinylmethyl)-2-pyridinamine
Hazard: Irritant
Formula:
C₁₁H₁₀BrN₃ |
MDL Number:
MFCD11123175 CAS Number: 1040008-79-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...